Assay Details
Target Gene Details
Entrez Gene ID: | 170755 |
Gene Name: | serum/glucocorticoid regulated kinase 3 |
Gene Aliases: |
2510015P22Rik, A330005P07Rik, Cisk, fy, fz |
Location: |
Chr.1:9798130-9902570 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk3 | NM_001037759.1 | 15 | 1367 | NP_001032848.1 |
| NM_133220.2 | 16 | 1486 | NP_573483.1 | |
| NM_177547.3 | 15 | 1379 | NP_808215.2 | |
| AF312007.1 | 16 | 1428 | AAG34115.1 | |
| AK028722.1 | 15 | 1722 | ||
| AK030314.1 | 15 | 1368 | BAC26895.1 | |
| AK031133.1 | 16 | 1467 | BAC27269.1 | |
| AK031328.1 | 15 | 1308 | BAC27349.1 | |
| AK083319.1 | 4 | 3454 | ||
| AK143836.1 | 16 | 1532 | ||
| AK146561.1 | 15 | 1322 | BAE27261.1 | |
| AK151181.1 | 16 | 1431 | BAE30182.1 | |
| AK151793.1 | BAE30694.1 | |||
| AK151867.1 | 16 | 1463 | BAE30755.1 | |
| AK151996.1 | 16 | 1630 | BAE30860.1 | |
| AK166414.1 | 15 | 1310 | ||
| AK166531.1 | 15 | 1328 | ||
| AK171120.1 | 15 | 1355 | BAE42262.1 | |
| AK171186.1 | 15 | 1355 | BAE42298.1 | |
| AK171678.1 | 15 | 1339 | ||
| AK171959.1 | 15 | 1416 | BAE42748.1 | |
| AK172323.1 | 15 | 1329 | BAE42945.1 | |
| AK186891.1 | ||||
| AK196812.1 | ||||
| BC018363.1 | 13 | 1017 | AAH18363.1 |
More Information
Set Membership: |
|

Genomic Map