Assay Details
Target Gene Details
Entrez Gene ID: | 76816 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
2700048G21Rik, 5730470G24Rik, Cccap |
Location: |
Chr.1:176814526-177020437 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdccag8 | NM_029756.3 | 2 | 436 | NP_084032.1 |
| AK012392.1 | 2 | 224 | BAB28209.1 | |
| AK017691.1 | 2 | 219 | BAB30877.1 | |
| AK051683.1 | 2 | 239 | BAC34719.1 | |
| AK132316.1 | 2 | 232 | BAE21100.1 | |
| AK133622.1 | 2 | 180 | BAE21753.1 | |
| AK136921.1 | 2 | 259 | BAE23170.1 | |
| BC016244.1 | 2 | 216 | ||
| BC025138.1 | 2 | 206 | AAH25138.1 | |
| CJ072722.1 | 2 | 288 |

Genomic Map