Assay Details
Target Gene Details
Entrez Gene ID: | 20344 |
Gene Name: | selectin, platelet |
Gene Aliases: |
CD62P, GMP-140, Grmp, LECAM3, PADGEM |
Location: |
Chr.1:164115264-164150026 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Selp | NM_011347.2 | 8 | 1403 | NP_035477.1 |
BC109158.1 | 7 | 1289 | AAI09159.1 | |
BC109159.1 | 7 | 1289 | AAI09160.1 | |
M72332.1 | 8 | 1403 | AAA37712.1 | |
M87861.1 | 7 | 1289 | AAA40008.1 |
