Assay Details
Target Gene Details
Entrez Gene ID: | 241197 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10 |
Gene Aliases: |
9830131G07, BB233602, Serpinb10-ps |
Location: |
Chr.1:107528994-107560735 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb10 | NM_001160307.1 | NP_001153779.1 | ||
| NM_198028.3 | 2 | 214 | NP_932145.3 | |
| AK036537.1 | 2 | 192 | ||
| AK052580.1 | ||||
| BC029736.1 | 2 | 155 | AAH29736.1 | |
| BC069938.1 | 2 | 174 | AAH69938.1 |
More Information
Set Membership: |
|

Genomic Map