Assay Details
Target Gene Details
Entrez Gene ID: | 241196 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13 |
Gene Aliases: |
5430417G24, HUR7, HURPIN, PI13, headpin |
Location: |
Chr.1:106980984-107001195 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb13 | NM_172852.3 | NP_766440.2 | ||
| AK030670.1 | BAC27072.1 | |||
| BC117990.1 | ||||
| BC117991.1 |
More Information
Set Membership: |
|

Genomic Map