Assay Details
Target Gene Details
Entrez Gene ID: | 21755 |
Gene Name: | protease, serine 39 |
Gene Aliases: |
Tesp1 |
Location: |
Chr.1:34498430-34503062 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss39 | NM_009355.2 | NP_033381.1 | ||
| AB008910.1 | BAA26132.1 | |||
| BC049616.1 | ||||
| BC115665.2 | ||||
| BC115666.2 |
More Information
Set Membership: |
|

Genomic Map