Assay Details
Target Gene Details
Entrez Gene ID: | 208777 |
Gene Name: | sushi, nidogen and EGF-like domains 1 |
Gene Aliases: |
6720455I24Rik, AI197264, AI642697, D430044C15Rik, SST3, Snep |
Location: |
Chr.1:93235815-93301065 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sned1 | NM_172463.4 | NP_766051.4 | ||
| AJ584850.1 | CAE48492.1 | |||
| AK086298.1 | ||||
| AY169783.1 | AAO41836.1 | |||
| BC145886.1 |

Genomic Map