Assay Details
Target Gene Details
Entrez Gene ID: | 20872 |
Gene Name: | serine/threonine kinase 16 |
Gene Aliases: |
EDPK, Krct, MPSK, PKL12, TSF-1 |
Location: |
Chr.1:75210829-75215606 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk16 | NM_001277992.1 | NP_001264921.1 | ||
| NM_011494.5 | NP_035624.3 | |||
| NR_102731.1 | ||||
| AA183397.1 | ||||
| AB017197.1 | BAA74457.1 | |||
| AB022179.1 | BAA89499.1 | |||
| AF062076.1 | AAD02811.1 | |||
| AF089869.1 | AAC98502.1 | |||
| AJ005790.1 | CAA06699.1 | |||
| AK030404.1 | BAC26947.1 | |||
| AK032162.1 | BAC27732.1 | |||
| AK149539.1 | BAE28946.1 | |||
| AK156554.1 | 4 | 633 | ||
| AK212366.1 | ||||
| BC005703.1 | ||||
| BC028999.1 | AAH28999.1 | |||
| CX200029.1 |
More Information
Set Membership: |
|

Genomic Map