Assay Details
Target Gene Details
Entrez Gene ID: | 22218 |
Gene Name: | SMT3 suppressor of mif two 3 homolog 1 (yeast) |
Gene Aliases: |
GMP1, PIC1, SENTRIN, SMT3, SMT3H3, SMTP3, SUMO-1, Smt3C, Ubl1 |
Location: |
Chr.1:59639434-59670834 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sumo1 | NM_009460.2 | 1 | 77 | NP_033486.1 |
AF033353.1 | 1 | 52 | AAC39959.1 | |
AK002536.1 | 1 | 47 | BAB22172.1 | |
AK011074.1 | 1 | 47 | BAB27379.1 | |
AK089081.1 | BAC40739.1 | |||
AK159366.1 | 1 | 48 | BAE35024.1 | |
BC082566.1 | 1 | 52 | AAH82566.1 | |
BC083158.1 | 1 | 50 | AAH83158.1 | |
BY242724.1 | 1 | 78 |
