Assay Details
Target Gene Details
Entrez Gene ID: | 226896 |
Gene Name: | transcription factor AP-2, delta |
Gene Aliases: |
Tcfap2d |
Location: |
Chr.1:19103022-19168592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfap2d | NM_153154.2 | NP_694794.1 | ||
| AF421891.1 | AAL16940.1 | |||
| AK132112.1 | ||||
| BC152324.1 | ||||
| BC152325.1 |
More Information
Set Membership: |
|

Genomic Map