Assay Details
Target Gene Details
Entrez Gene ID: | 277328 |
Gene Name: | transient receptor potential cation channel, subfamily A, member 1 |
Gene Aliases: |
Anktm1, TRPA1b |
Location: |
Chr.1:14872648-14919014 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpa1 | NM_177781.4 | NP_808449.1 | ||
| AK045771.1 | BAC32487.1 | |||
| AY231177.1 | AAO43183.1 | |||
| BC120563.1 | ||||
| BC131963.1 | ||||
| KF460461.1 |
More Information
Set Membership: |
|

Genomic Map