Assay Details
Target Gene Details
Entrez Gene ID: | 22145 |
Gene Name: | tubulin, alpha 4A |
Gene Aliases: |
M[a]4, Tuba4 |
Location: |
Chr.1:75214974-75219860 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba4a | NM_009447.3 | NP_033473.1 | ||
| AK002427.1 | BAB22094.1 | |||
| AK078355.1 | BAC37234.1 | |||
| AK158934.1 | BAE34732.1 | |||
| BC019959.1 | AAH19959.1 | |||
| M13444.1 | AAA40502.1 |
More Information
Set Membership: |
|

Genomic Map