Assay Details
Target Gene Details
Entrez Gene ID: | 319266 |
Gene Name: | RIKEN cDNA A130010J15 gene |
Gene Aliases: |
- |
Location: |
Chr.1:193173469-193177832 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A130010J15Rik | NM_001160359.1 | 1 | 97 | NP_001153831.1 |
| NM_001160360.1 | NP_001153832.1 | |||
| NM_181048.3 | 1 | 97 | NP_851391.1 | |
| AK037364.1 | 1 | 94 | BAC29794.1 | |
| AK047686.1 | BAC33124.1 | |||
| AK077339.1 | BAC36759.1 | |||
| AK084448.1 | BAC39186.1 | |||
| BB858416.1 | 1 | 91 | ||
| BC055955.1 | AAH55955.1 | |||
| BY011244.1 | 1 | 97 |

Genomic Map