Assay Details
Target Gene Details
Entrez Gene ID: | 213452 |
Gene Name: | dual serine/threonine and tyrosine protein kinase |
Gene Aliases: |
A930019K20Rik, C430014H23Rik, C820013G01, Ripk5 |
Location: |
Chr.1:132417181-132466959 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dstyk | NM_172516.4 | 4 | 1625 | NP_766104.2 |
| AK083696.1 | 4 | 1523 | ||
| AK132119.1 | 4 | 1500 | ||
| AK145449.1 | 4 | 1470 | BAE26444.1 | |
| AK172954.1 | 4 | 1471 | BAD32232.1 | |
| AY208852.1 | 4 | 1472 | AAP42420.1 | |
| AY429676.1 | 4 | 1511 | AAS55392.1 | |
| BC089380.1 | 1 | 42 | AAH89380.1 | |
| BC130226.1 | 4 | 1501 |
More Information
Set Membership: |
|

Genomic Map