Assay Details
Target Gene Details
Entrez Gene ID: | 26754 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana) |
Gene Aliases: |
AI303502, CSN5, Jab1, Mov34, Sgn5 |
Location: |
Chr.1:10024600-10038159 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops5 | NM_001277101.1 | NP_001264030.1 | ||
| NM_013715.2 | NP_038743.1 | |||
| NR_102282.1 | 1 | 461 | ||
| AF065386.1 | AAC17179.1 | |||
| AF068223.1 | AAF61318.1 | |||
| AK002761.1 | ||||
| AK005113.1 | 1 | 248 | ||
| AK005728.1 | ||||
| AK012499.1 | BAB28282.1 | |||
| AK133073.1 | BAE21497.1 | |||
| AK146271.1 | BAE27030.1 | |||
| AK151492.1 | BAE30445.1 | |||
| BC046753.1 | AAH46753.1 | |||
| CX240457.1 | ||||
| DV056728.1 | 1 | 260 | ||
| U70736.1 | AAD03470.1 |
More Information
Set Membership: |
|

Genomic Map