Assay Details
Target Gene Details
Entrez Gene ID: | 14270 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2 |
Gene Aliases: |
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3 |
Location: |
Chr.1:131285251-131527369 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srgap2 | NM_001081011.2 | 8 | 1665 | NP_001074480.2 |
| BC151081.1 | 7 | 1283 | ||
| BC151082.1 | 7 | 1283 | ||
| BC158055.1 | 7 | 1281 | ||
| BC172152.1 | 7 | 1283 | ||
| BE653295.1 | 1 | 54 |
More Information
Set Membership: |
|

Genomic Map