Assay Details
Target Gene Details
Entrez Gene ID: | 20872 |
Gene Name: | serine/threonine kinase 16 |
Gene Aliases: |
EDPK, Krct, MPSK, PKL12, TSF-1 |
Location: |
Chr.1:75210829-75215606 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk16 | NM_001277992.1 | 3 | 524 | NP_001264921.1 |
| NM_011494.5 | 3 | 552 | NP_035624.3 | |
| NR_102731.1 | 3 | 552 | ||
| AA183397.1 | ||||
| AB017197.1 | 3 | 500 | BAA74457.1 | |
| AB022179.1 | 3 | 482 | BAA89499.1 | |
| AF062076.1 | 2 | 239 | AAD02811.1 | |
| AF089869.1 | 3 | 529 | AAC98502.1 | |
| AJ005790.1 | 3 | 483 | CAA06699.1 | |
| AK030404.1 | 3 | 527 | BAC26947.1 | |
| AK032162.1 | 3 | 543 | BAC27732.1 | |
| AK149539.1 | 3 | 525 | BAE28946.1 | |
| AK156554.1 | 3 | 455 | ||
| BC005703.1 | 3 | 511 | ||
| BC028999.1 | 3 | 511 | AAH28999.1 | |
| CX200029.1 | 3 | 499 | ||
| DQ544387.1 |
More Information
Set Membership: |
|

Genomic Map