Assay Details
Target Gene Details
Entrez Gene ID: | 98432 |
Gene Name: | PH domain and leucine rich repeat protein phosphatase 1 |
Gene Aliases: |
AI836256, Phlpp, Plekhe1, SCOP, mKIAA0606 |
Location: |
Chr.1:106171869-106394245 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phlpp1 | NM_133821.3 | 16 | 3825 | NP_598582.3 |
| AB093251.1 | 16 | 2584 | BAC41435.1 | |
| BC024670.1 | 4 | 453 | AAH24670.1 | |
| BC059254.1 | 16 | 2485 | AAH59254.1 |
More Information
Set Membership: |
|

Genomic Map