Assay Details
Target Gene Details
Entrez Gene ID: | 269152 |
Gene Name: | kinesin family member 26B |
Gene Aliases: |
4832420M10, BC056349, D230039L06Rik |
Location: |
Chr.1:178525741-178939200 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif26b | NM_001161665.1 | 13 | 6335 | NP_001155137.1 |
| AB355846.1 | 13 | 6335 | ||
| AK076435.1 | 2 | 2080 | BAC36343.1 | |
| BC056349.1 | 8 | 4418 | AAH56349.1 |
More Information
Set Membership: |
|

Genomic Map