Assay Details
Target Gene Details
Entrez Gene ID: | 240756 |
Gene Name: | kelch-like 12 |
Gene Aliases: |
C3ip1 |
Location: |
Chr.1:134455495-134490875 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl12 | NM_153128.2 | 2 | 268 | NP_694768.1 |
| AK034170.1 | 2 | 256 | BAC28614.1 | |
| AK165280.1 | 2 | 246 | ||
| BC034514.1 | 2 | 268 | AAH34514.1 |
More Information
Set Membership: |
|

Genomic Map