Assay Details
Target Gene Details
Entrez Gene ID: | 19253 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 18 |
Gene Aliases: |
FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1 |
Location: |
Chr.1:34459746-34473784 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpn18 | NM_011206.2 | NP_035336.2 | ||
AK144353.1 | BAE25845.1 | |||
BC008512.1 | AAH08512.1 | |||
CD740968.1 | ||||
U35124.1 | AAB82736.1 | |||
U49853.1 | AAB18623.1 | |||
U52523.1 | AAC52991.1 |
