Assay Details
Target Gene Details
Entrez Gene ID: | 19731 |
Gene Name: | ral guanine nucleotide dissociation stimulator,-like 1 |
Gene Aliases: |
Rgl, mKIAA0959 |
Location: |
Chr.1:152516768-152766351 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgl1 | NM_016846.3 | 7 | 944 | NP_058542.2 |
| AK083967.1 | 1 | 517 | ||
| AK144218.1 | 7 | 1019 | BAE25780.1 | |
| AK169109.1 | 7 | 996 | BAE40891.1 | |
| AK169180.1 | 7 | 997 | BAE40958.1 | |
| BC017678.1 | 7 | 944 | AAH17678.1 | |
| U14103.1 | 7 | 1002 | AAA64950.1 |
More Information
Set Membership: |
|

Genomic Map