Assay Details
Target Gene Details
Entrez Gene ID: | 76816 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
2700048G21Rik, 5730470G24Rik, Cccap |
Location: |
Chr.1:176814526-177020437 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdccag8 | NM_029756.3 | 1 | 276 | NP_084032.1 |
AK012392.1 | 1 | 64 | BAB28209.1 | |
AK017691.1 | 1 | 59 | BAB30877.1 | |
AK051683.1 | 1 | 79 | BAC34719.1 | |
AK132316.1 | 1 | 72 | BAE21100.1 | |
AK133622.1 | BAE21753.1 | |||
AK136921.1 | 1 | 99 | BAE23170.1 | |
BC016244.1 | 1 | 56 | ||
BC025138.1 | 1 | 46 | AAH25138.1 | |
CJ072722.1 | 1 | 128 |
