Assay Details
Target Gene Details
Entrez Gene ID: | 67273 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10 |
Gene Aliases: |
2900053E13Rik |
Location: |
Chr.1:92439708-92473758 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa10 | NM_024197.1 | 3 | 381 | NP_077159.1 |
| AK005339.1 | 3 | 417 | BAB23961.2 | |
| AK013675.1 | 3 | 425 | ||
| AK145979.1 | 3 | 437 | BAE26801.1 | |
| AK177419.1 | 2 | 235 | ||
| AK184477.1 | 2 | 235 | ||
| AK185477.1 | 2 | 234 | ||
| BC003439.1 | 3 | 381 | AAH03439.1 |
More Information
Set Membership: |
|

Genomic Map