Assay Details
Target Gene Details
Entrez Gene ID: | 68295 |
Gene Name: | AAR2 splicing factor homolog (S. cerevisiae) |
Gene Aliases: |
0610011L14Rik |
Location: |
Chr.2:156547558-156568972 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aar2 | NM_001164818.1 | NP_001158290.1 | ||
| NM_026661.4 | NP_080937.3 | |||
| AK018741.1 | BAB31382.1 | |||
| AK028890.1 | BAC26177.1 | |||
| AK049107.1 | ||||
| AK166203.1 | BAE38626.1 | |||
| AK169526.1 | BAE41210.1 | |||
| AK180344.1 | ||||
| AK185720.1 | ||||
| AK187378.1 | ||||
| AK189523.1 | ||||
| AK199836.1 | ||||
| AK208290.1 | ||||
| AK210218.1 | ||||
| BC019988.1 | AAH19988.1 | |||
| BC020112.1 | AAH20112.1 |
More Information
Set Membership: |
|

Genomic Map