Assay Details
Target Gene Details
Entrez Gene ID: | 67388 |
Gene Name: | RIKEN cDNA 1110008F13 gene |
Gene Aliases: |
AI316789 |
Location: |
Chr.2:156863122-156873563 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008F13Rik | NM_026124.3 | 2 | 353 | NP_080400.1 |
| AK002452.1 | 2 | 311 | BAB22111.1 | |
| AK003568.1 | 2 | 348 | BAB22864.1 | |
| AK031002.1 | 2 | 331 | ||
| AK078178.1 | 2 | 349 | ||
| AK179243.1 | 2 | 74 | ||
| AK186989.1 | 2 | 74 | ||
| BC019445.1 | 2 | 291 | AAH19445.1 | |
| BY165552.1 |

Genomic Map