Assay Details
Target Gene Details
Entrez Gene ID: | 73358 |
Gene Name: | RIKEN cDNA 1700054M17 gene |
Gene Aliases: |
- |
Location: |
Chr.2:118304914-118308166 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700054M17Rik | NR_045919.1 | |||
| AK006785.1 |
Target Gene Details
Entrez Gene ID: | 381413 |
Gene Name: | G protein-coupled receptor 176 |
Gene Aliases: |
Gm1012 |
Location: |
Chr.2:118277096-118373419 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr176 | NM_201367.3 | NP_958755.1 | ||
| BC052037.1 | AAH52037.1 |
More Information
Set Membership: |
|

Genomic Map