Assay Details
Target Gene Details
Entrez Gene ID: | 71532 |
Gene Name: | family with sequence similarity 217, member B |
Gene Aliases: |
9030418K01Rik, AI851938 |
Location: |
Chr.2:178414534-178422181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam217b | NM_001081289.1 | NP_001074758.1 | ||
| AK155442.1 |
More Information
Set Membership: |
|

Genomic Map