Assay Details
Target Gene Details
Entrez Gene ID: | 74702 |
Gene Name: | RIKEN cDNA 4930517E11 gene |
Gene Aliases: |
- |
Location: |
Chr.2:124123636-124129956 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930517E11Rik | NR_040611.1 | |||
| AK015809.1 |
Target Gene Details
Entrez Gene ID: | 214968 |
Gene Name: | sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D |
Gene Aliases: |
1110067B02Rik, AA409156, D330011G23, mKIAA1479 |
Location: |
Chr.2:124089969-124667789 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema6d | NM_001290997.1 | NP_001277926.1 | ||
| AK122515.1 | BAC65797.1 | |||
| BY242475.1 |
More Information
Set Membership: |
|

Genomic Map