Assay Details
Target Gene Details
Entrez Gene ID: | 78774 |
Gene Name: | cilia and flagella associated protein 61 |
Gene Aliases: |
4930529M08Rik, BB178539, BC024760 |
Location: |
Chr.2:145934784-145964226 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfap61 | NM_175280.3 | NP_780489.2 | ||
| AK038772.1 | ||||
| BC057361.1 | AAH57361.1 | |||
| BC058733.1 | AAH58733.1 | |||
| BC107030.1 | AAI07031.1 | |||
| BC107031.1 | AAI07032.1 |
More Information
Set Membership: |
|

Genomic Map