Assay Details
Target Gene Details
Entrez Gene ID: | 241694 |
Gene Name: | Ral GTPase activating protein, alpha subunit 2 (catalytic) |
Gene Aliases: |
A230067G21Rik, AS250, BC053994, RGC2, pp250 |
Location: |
Chr.2:146239124-146512222 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgapa2 | NM_001033348.3 | 31 | 4048 | NP_001028520.2 |
| BC053994.1 | 9 | 909 | AAH53994.1 | |
| BC096528.1 | 3 | 246 | AAH96528.1 | |
| CJ114686.1 |
More Information
Set Membership: |
|

Genomic Map