Assay Details
Target Gene Details
Entrez Gene ID: | 77806 |
Gene Name: | RIKEN cDNA A930019D19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:146259286-146267160 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A930019D19Rik | NR_040619.1 | |||
| AK020875.1 |
Target Gene Details
Entrez Gene ID: | 241694 |
Gene Name: | Ral GTPase activating protein, alpha subunit 2 (catalytic) |
Gene Aliases: |
A230067G21Rik, AS250, BC053994, RGC2, pp250 |
Location: |
Chr.2:146239124-146512222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ralgapa2 | NM_001033348.3 | NP_001028520.2 | ||
| AK042986.1 | ||||
| AK173140.1 | BAD32418.1 | |||
| BC053994.1 | AAH53994.1 | |||
| BC096528.1 | AAH96528.1 |
More Information
Set Membership: |
|

Genomic Map