Assay Details
Target Gene Details
Entrez Gene ID: | 75763 |
Gene Name: | DDB1 and CUL4 associated factor 17 |
Gene Aliases: |
2810055O12Rik, 4833418A01Rik, A030004A10Rik, A930009G19Rik, AI448937 |
Location: |
Chr.2:71055742-71114514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf17 | NM_001165980.1 | NP_001159452.1 | ||
| NM_001165981.1 | NP_001159453.1 | |||
| NM_198005.2 | NP_932122.2 | |||
| AK014717.1 | ||||
| AK081839.1 | ||||
| AK160679.1 | BAE35954.1 | |||
| BB772704.1 | ||||
| BC031546.1 | AAH31546.1 |

Genomic Map