Assay Details
Target Gene Details
Entrez Gene ID: | 74614 |
Gene Name: | osteoclast stimulatory transmembrane protein |
Gene Aliases: |
4833422F24Rik, OC-STAMP |
Location: |
Chr.2:165395450-165400394 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ocstamp | NM_029021.1 | NP_083297.1 | ||
| AK014751.1 | BAB29531.1 | |||
| AK042271.1 | BAC31210.1 | |||
| BC020160.1 | AAH20160.1 |

Genomic Map