Assay Details
Target Gene Details
Entrez Gene ID: | 66766 |
Gene Name: | transmembrane 239 |
Gene Aliases: |
4933425O20Rik |
Location: |
Chr.2:130406522-130407794 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem239 | NM_025753.3 | NP_080029.1 | ||
| AK006188.1 | BAB24448.1 | |||
| AK016923.1 | BAC25500.1 | |||
| BC049728.1 | AAH49728.1 |

Genomic Map