Assay Details
Target Gene Details
Entrez Gene ID: | 228602 |
Gene Name: | RIKEN cDNA 4930402H24 gene |
Gene Aliases: |
2810487F15Rik, AI413763, C78926 |
Location: |
Chr.2:130706200-130840807 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930402H24Rik | NM_029432.1 | 22 | 1926 | NP_083708.1 |
| AK133294.1 | 22 | 2068 | BAE21597.1 | |
| AK143611.1 | 22 | 1951 | BAE25462.1 | |
| AK158367.1 | 22 | 1951 | BAE34475.1 | |
| AK207494.1 | 1 | 54 | ||
| BC052447.1 | 22 | 1926 | AAH52447.2 |

Genomic Map