Assay Details
Target Gene Details
Entrez Gene ID: | 329540 |
Gene Name: | nucleolar protein 4-like |
Gene Aliases: |
4931410H15, 8430427H17Rik, AI429157, Gm1636 |
Location: |
Chr.2:153407461-153529971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol4l | NM_001001986.2 | NP_001001986.2 | ||
| NM_001134300.2 | NP_001127772.1 | |||
| AK139459.1 | BAE24020.1 | |||
| AK147524.1 | BAE27972.1 | |||
| AK157863.1 | BAE34240.1 | |||
| BC075648.1 | AAH75648.1 | |||
| BC079552.1 | AAH79552.1 | |||
| BC094234.1 | AAH94234.1 |

Genomic Map