Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abl1 | NM_001112703.2 | 2 | 1489 | NP_001106174.1 |
NM_001283045.1 | 2 | 366 | NP_001269974.1 | |
NM_001283046.1 | 3 | 1694 | NP_001269975.1 | |
NM_001283047.1 | 3 | 1633 | NP_001269976.1 | |
NM_009594.4 | 2 | 260 | NP_033724.2 | |
AK088252.1 | 3 | 1619 | BAE43394.1 | |
AK090095.1 | 2 | 218 | BAC41088.1 | |
AK166028.1 | 2 | 1478 | ||
AK166210.1 | 3 | 1728 | BAE38629.1 | |
AK194745.1 | 2 | 197 | ||
BC059260.1 | 2 | 287 | AAH59260.1 | |
BC103770.1 | 2 | 713 | AAI03771.1 | |
BY152805.1 | 2 | 257 | ||
CJ099623.1 | 2 | 211 | ||
CJ101520.1 | 2 | 260 | ||
CJ128107.1 | 2 | 255 | ||
CN663033.1 | ||||
J02995.1 | 2 | 213 | AAA88241.1 | |
K03228.1 | 2 | 255 | AAA37135.1 | |
M12263.1 | 3 | 620 | AAA37136.1 | |
M12264.1 | 2 | 366 | AAA37137.1 | |
M12265.1 | 2 | 415 | AAA37138.1 |
