Assay Details
Target Gene Details
Entrez Gene ID: | 80908 |
Gene Name: | ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase) |
Gene Aliases: |
NAGAT |
Location: |
Chr.2:26842496-26864995 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abo | NM_001290444.1 | NP_001277373.1 | ||
| NM_030718.5 | NP_109643.3 | |||
| AB041039.1 | BAB20560.1 | |||
| AK033786.1 | BAC28473.1 | |||
| AK035261.1 | BAC29005.1 | |||
| BC116759.1 | ||||
| BC116761.1 | ||||
| DV046627.1 |
More Information
Set Membership: |
|

Genomic Map