Assay Details
Target Gene Details
Entrez Gene ID: | 228998 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 1 |
Gene Aliases: |
AI115377, Arf1gap |
Location: |
Chr.2:180967225-180982524 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap1 | NM_001177706.1 | 1 | 444 | NP_001171177.1 |
| NM_001177707.1 | 2 | 315 | NP_001171178.1 | |
| NM_001177708.1 | NP_001171179.1 | |||
| NM_001177709.1 | NP_001171180.1 | |||
| NM_001177710.1 | NP_001171181.1 | |||
| NM_145760.3 | 2 | 247 | NP_665703.2 | |
| AJ401461.1 | 2 | 205 | CAC18721.1 | |
| AK030295.1 | 2 | 203 | BAC26883.1 | |
| AK030520.1 | 1 | 446 | BAC27002.1 | |
| AK052985.1 | 1 | 462 | BAC35231.1 | |
| AK089321.1 | 1 | 466 | ||
| AK134402.1 | 2 | 257 | BAE22128.1 | |
| AK145879.1 | 2 | 231 | BAE26720.1 | |
| AK168007.1 | 2 | 228 | BAE39994.1 | |
| AK168602.1 | BAE40469.1 | |||
| AK219899.1 | 1 | 51 | ||
| BC016228.1 | ||||
| BC052922.1 | ||||
| BC059817.1 | AAH59817.1 | |||
| CJ149044.1 | ||||
| CJ150232.1 |
More Information
Set Membership: |
|

Genomic Map