Assay Details
Target Gene Details
Entrez Gene ID: | 228998 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 1 |
Gene Aliases: |
AI115377, Arf1gap |
Location: |
Chr.2:180967225-180982524 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap1 | NM_001177706.1 | NP_001171177.1 | ||
| NM_001177707.1 | NP_001171178.1 | |||
| NM_001177708.1 | NP_001171179.1 | |||
| NM_001177709.1 | NP_001171180.1 | |||
| NM_001177710.1 | NP_001171181.1 | |||
| NM_145760.3 | NP_665703.2 | |||
| AJ401461.1 | CAC18721.1 | |||
| AK030295.1 | BAC26883.1 | |||
| AK030520.1 | BAC27002.1 | |||
| AK134402.1 | BAE22128.1 | |||
| AK145879.1 | BAE26720.1 | |||
| AK168007.1 | BAE39994.1 | |||
| AK168602.1 | BAE40469.1 | |||
| AK219899.1 | ||||
| BC016228.1 | ||||
| BC052922.1 | ||||
| BC059817.1 | AAH59817.1 | |||
| CJ149044.1 | ||||
| CJ150232.1 |
More Information
Set Membership: |
|

Genomic Map