Assay Details
Target Gene Details
Entrez Gene ID: | 76688 |
Gene Name: | ADP-ribosylation factor related protein 1 |
Gene Aliases: |
1500006I01Rik, AI480700 |
Location: |
Chr.2:181357690-181365404 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfrp1 | NM_001165991.1 | 1 | 127 | NP_001159463.1 |
| NM_001165992.1 | NP_001159464.1 | |||
| NM_001165995.1 | NP_001159467.1 | |||
| NM_029702.4 | NP_083978.3 | |||
| NR_028585.1 | ||||
| AJ413953.1 | 1 | 74 | CAC88692.1 | |
| AK005174.1 | BAB23861.1 | |||
| AK044759.1 | BAC32069.1 | |||
| AK050136.1 | 1 | 58 | ||
| BC010713.1 | AAH10713.1 | |||
| BC046782.1 | AAH46782.1 | |||
| BC046786.1 | AAH46786.1 | |||
| CA539216.1 |
Target Gene Details
Entrez Gene ID: | 229007 |
Gene Name: | zinc finger, CCCH-type with G patch domain |
Gene Aliases: |
1500006I01Rik, BC021513 |
Location: |
Chr.2:181364931-181380793 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zgpat | NM_144894.3 | 1 | 348 | NP_659143.1 |
| AK041491.1 | 1 | 351 | BAC30962.1 | |
| BY287149.1 | 1 | 53 |

Genomic Map