Assay Details
Target Gene Details
Entrez Gene ID: | 279028 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 13 |
Gene Aliases: |
ADAM-TS13, ADAMTS-13, Gm710, vWF-CP |
Location: |
Chr.2:26973416-27009628 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts13 | NM_001001322.2 | 20 | 2696 | NP_001001322.1 |
| NM_001290463.1 | 20 | 2612 | NP_001277392.1 | |
| NM_001290464.1 | 20 | 2696 | NP_001277393.1 | |
| NM_001290465.1 | 19 | 2669 | NP_001277394.1 | |
| AB071302.2 | 20 | 2696 | BAC55159.2 | |
| AB112362.1 | 20 | 2575 | BAD18090.1 | |
| AK157459.1 | 20 | 2537 | ||
| EU034706.1 | 19 | 2548 |
More Information
Set Membership: |
|

Genomic Map