Assay Details
Target Gene Details
Entrez Gene ID: | 99152 |
Gene Name: | anaphase promoting complex subunit 2 |
Gene Aliases: |
9230107K09Rik, AL024279, Apc2, Emi4, Imi4, mKIAA1406 |
Location: |
Chr.2:25272466-25285916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc2 | NM_175300.4 | NP_780509.2 | ||
| AK033784.1 | BAC28472.1 | |||
| AK129351.1 | BAC98161.1 | |||
| AK154465.1 | BAE32605.1 | |||
| AK170676.1 | BAE41951.1 | |||
| AK186163.1 | ||||
| AK186633.1 | ||||
| AK196194.1 | ||||
| BC027351.1 | AAH27351.1 | |||
| BC137583.1 |

Genomic Map