Assay Details
Target Gene Details
Entrez Gene ID: | 56436 |
Gene Name: | adhesion regulating molecule 1 |
Gene Aliases: |
1110063P18Rik, 2510006J17Rik, AA408205, ARM-1, AU043535, Arm1, Gp110, Rpn13 |
Location: |
Chr.2:180171490-180176286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adrm1 | NM_019822.3 | NP_062796.2 | ||
AF225959.1 | AAF33401.1 | |||
AK010919.1 | ||||
AK075674.1 | BAC35889.1 | |||
AK082084.1 | ||||
AK143843.1 | 3 | 1362 | ||
AK145474.1 | BAE26457.1 | |||
AK145792.1 | BAE26653.1 | |||
AK150091.1 | ||||
AK166878.1 | BAE39089.1 | |||
AK167085.1 | 1 | 1774 | ||
BC008974.1 | AAH08974.1 | |||
BC019746.1 | AAH19746.1 | |||
BC031517.1 | AAH31517.1 |
