Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf2 | NM_001025093.2 | 13 | 1458 | NP_001020264.1 |
NM_001284369.1 | 11 | 1298 | NP_001271298.1 | |
NM_001284370.1 | 13 | 1458 | NP_001271299.1 | |
NM_001284371.1 | 12 | 1395 | NP_001271300.1 | |
NM_001284372.1 | 11 | 1164 | NP_001271301.1 | |
NM_001284373.1 | 10 | 1101 | NP_001271302.1 | |
NM_001284374.1 | 10 | 1183 | NP_001271303.1 | |
NM_001284376.1 | 13 | 1340 | NP_001271305.1 | |
NM_009715.3 | 12 | 1361 | NP_033845.1 | |
AF483482.1 | 9 | 1035 | AAL90756.1 | |
AF483483.1 | 9 | 1035 | AAL90757.1 | |
AK035438.1 | 13 | 1414 | BAC29065.1 | |
AK046184.1 | 2 | 219 | BAC32623.1 | |
AK051591.1 | 12 | 1351 | BAC34684.1 | |
AK139515.1 | 11 | 1214 | ||
AK144977.1 | 11 | 1138 | ||
AK145887.1 | 11 | 1068 | BAE26726.1 | |
AK146839.1 | 12 | 1307 | ||
AY902311.1 | 8 | 921 | AAX83926.1 | |
BC042210.1 | 10 | 1134 | AAH42210.1 | |
BC079883.1 | 12 | 1283 | AAH79883.1 | |
BC082596.1 | 10 | 1047 | AAH82596.1 | |
M31629.1 | 5 | 513 | AAA39780.1 | |
M77167.1 | 7 | 807 | AAA40394.1 | |
U46026.1 | 2 | 303 | AAB64016.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
