Assay Details
Target Gene Details
Entrez Gene ID: | 228482 |
Gene Name: | Rho GTPase activating protein 11A |
Gene Aliases: |
6530401L14Rik, mKIAA0013 |
Location: |
Chr.2:113831492-113848661 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap11a | NM_181416.3 | NP_852081.2 | ||
| AK032639.1 | BAC27965.1 | |||
| AK137111.1 |
More Information
Set Membership: |
|

Genomic Map