Assay Details
Target Gene Details
Entrez Gene ID: | 65103 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 6 |
Gene Aliases: |
2310057C01Rik, 2610529A11Rik, Aip-6 |
Location: |
Chr.2:53192084-53219221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl6ip6 | NM_022989.4 | NP_075365.3 | ||
| AK011070.1 | BAB27377.1 | |||
| AK016267.1 | BAB30169.1 | |||
| AK032740.1 | BAC28004.1 | |||
| AK035521.1 | BAC29090.1 | |||
| AK052010.1 | BAC34831.1 | |||
| AK082824.1 | BAC38638.1 | |||
| AK159990.1 | BAE35540.1 | |||
| BC019550.1 | AAH19550.1 | |||
| BC157980.1 |
More Information
Set Membership: |
|

Genomic Map