Assay Details
Target Gene Details
Entrez Gene ID: | 12048 |
Gene Name: | BCL2-like 1 |
Gene Aliases: |
Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 |
Location: |
Chr.2:152758732-152832581 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l1 | NM_009743.5 | NP_033873.3 | ||
| AK150028.1 | ||||
| BB856100.1 | ||||
| BC089016.1 | AAH89016.1 |
More Information
Set Membership: |
|

Genomic Map