Assay Details
Target Gene Details
Entrez Gene ID: | 403171 |
Gene Name: | barrier to autointegration factor 2 |
Gene Aliases: |
4930517K23Rik, Baf-L, Baf-like, Gm115 |
Location: |
Chr.2:144033102-144073979 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Banf2 | NM_001044750.1 | NP_001038215.1 | ||
| NM_207275.1 | NP_997158.1 | |||
| AK076863.1 | BAC36510.1 |

Genomic Map